Almandine Garnet
Fournisseur: Avantor
Certificats
A propos de cet article
Exemplary display of hexoctahedral crystal shape.
- Available in multiple sizes
Classroom sized pieces are ideal for students to use for study and identification. Specimen is on average 0.5×0.5". Display specimens are great for lectures, demonstrations, and display. Specimen is on average 1×1".
Customers Who Bought This Also Bought