Allium, Epidermis Slide
Catalog # 470177-456
Supplier: Avantor
Specifications
- Description:Allium, Epidermis
- Cat. no.:470177-456
- Stain:Fuchsin and fast green
- Supplier No.:WARD470177-456
- Orientation:Whole mount
Specifications
About this item
Explore simple plant cell structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Sourced from an onion bulb. Shows simple plant cells.
Customers Who Bought This Also Bought