ContactUsLinkComponent
Allium, Epidermis Slide
Allium, Epidermis Slide
Catalog # 470177-456
Supplier:  Avantor
ONION (ALLIUM) BULB EPIDERMIS WM
Allium, Epidermis Slide
Catalog # 470177-456
Supplier:  Avantor
Specifications
  • Description:
    Allium, Epidermis
  • Cat. no.:
    470177-456
  • Stain:
    Fuchsin and fast green
  • Supplier No.:
    WARD470177-456
  • Orientation:
    Whole mount
Specifications
About this item
Explore simple plant cell structure
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Sourced from an onion bulb. Shows simple plant cells.
Customers Who Bought This Also Bought