Alligator Test Clips
# de catalogue 470176-120
Fournisseur: TOP LAB CORP.
Spécifications
- Description:Alligator Test Clips
- Cat. no.:470176-120
- Supplier No.:CTA-004
Spécifications
A propos de cet article
Long nosed clips specially suitable for electrical connections.
- Constructed of plated steel
They have a 5 A capacity and a jaw spread of 9.5 mm.
Packaging: Pack of 12.
Customers Who Bought This Also Bought