Alligator Clip/Banana Plug Cords
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Easily connect a banana plug to a bare connector.
Cords, 61 cm long, with an alligator clip at one end and a two way banana plug at the other. The alligator clip has a protective vinyl plastic sheath. The flexible plastic coated wire has a rating of 15 A, AC.
- Flexible plastic coated wire
- 15 Amp rating
Cords, 61 cm long, with an alligator clip at one end and a two way banana plug at the other. The alligator clip has a protective vinyl plastic sheath. The flexible plastic coated wire has a rating of 15 A, AC.
Customers Who Bought This Also Bought