Advanced Earth Science Collection
# de catalogue 470112-324
Fournisseur: Geoscience Industries
Spécifications
- Description:Advanced Earth Science Collection
- No. of Samples:105
- Cat. no.:470112-324
- Supplier No.:1202 + PKG
Spécifications
A propos de cet article
The Advanced Earth Science Collection features a large, detailed collection of specimens to provide a better understanding of earth’s composition.
- Study Guide Included
- Number of Samples: 105
- Many Rock and Mineral Properties Included
- Length: 1"
- Width: 1"
This set contains a total of 105 specimens group packaged in nine separate boxes.
The specimen size is approximately 1 x 1".
Ordering information: This kit includes cleavage and fracture; streak, color and luster; rock-forming minerals, Parts 1 and 2; igneous rocks; metamorphic rocks; sedimentary rocks; and common fossils. Nine specimens illustrating the hardness scale, a streak plate, magnet, magnifier, rock classification tables, a geologic time chart, a key sheet, and study guide are also included.
Customers Who Bought This Also Bought