Adult Scalp Slide
# de catalogue 470176-996
Fournisseur: Avantor
Spécifications
- Description:Adult Scalp
- Cat. no.:470176-996
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470176-996
- Orientation:Cross section
Spécifications
A propos de cet article
Examine the epidermis of the head
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Hair follicles at different levels.
Customers Who Bought This Also Bought