Adjustable Aluminum Tuning Fork
# de catalogue 470104-596
Fournisseur: RIVERBANK LABORATORIES INC.
Spécifications
- Description:Adjustable Tuning Fork
- Cat. no.:470104-596
- Supplier No.:N
Spécifications
A propos de cet article
This aluminum tuning fork has sliding weights on each tine.
- High-Quality
- Adjustable
- Sliding Weights on Each Tine
Index lines and notes indicate settings from C (128Hz) to B (240Hz).
This high-quality tuning fork is adjustable
Customers Who Bought This Also Bought