Adhesive Clips
# de catalogue 470020-452
Fournisseur: FORENSICS SOURCE
Spécifications
- Description:Adhesive Clips
- Cat. no.:470020-452
- Supplier No.:ADCL-1
Spécifications
A propos de cet article
Use clips to attach string to any vertical surface for ballistics determinations.
- Removable and reusable
- Package of 10
The colorless adhesive wax backing on these clips will never dry out, so you can remove and reuse them repeatedly.
Customers Who Bought This Also Bought