Ward's® Live Actinosphaerium Sp. Culture
# de catalogue 470176-356
Fournisseur: Ward's Science
Spécifications
- Description:Actinosphaerium (small)
- Storage Conditions:Room temp, loose cap
- No. of Students:30
- Cat. no.:470176-356
- Packaging:Jar
- Media:Soil water medium
- Supplier No.:WARD470176-356
Spécifications
A propos de cet article
With stiff 'arms' radiating in all directions, these microscopic organisms look like sea urchins
- Large protist
- Excellent example of a Heliozoan, or "sun animal".
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought