Acrylic Refraction Cube
# de catalogue 470149-494
Fournisseur: VWR International
Spécifications
- Description:Acrylic Refraction Cube
- Cat. no.:470149-494
- Supplier No.:P30347
Spécifications
A propos de cet article
Ideal for Index of Refraction and Total Internal Reflection Experiments
The 5cm faces on this acrylic cube are transparent and highly polished for clarity. The cube has an index of refraction of about 1.5.
Customers Who Bought This Also Bought