Acrylic Pump Plate
# de catalogue 470121-116
Fournisseur: Fischer Technical
Spécifications
- Description:Acrylic pump plate
- Supplier No.:AP-001
Spécifications
A propos de cet article
The durable acrylic pump plate accommodates bell jars up to 9" Ø.
- Durable
- Vacuum release valve included
- Rubber gasket included
- Barbed-hose connection included
- Plate fits bell jars up to 9" Ø.
It includes a rubber gasket for a tight seal.
The plate also includes a vacuum release valve and 3/8" barbed-hose connection.
Customers Who Bought This Also Bought