Acrylic Prism Set
# de catalogue 470148-722
Fournisseur: VWR International
Spécifications
- Description:Acrylic Prism Set
- Cat. no.:470148-722
- Supplier No.:P30277
Spécifications
A propos de cet article
A must for your optics lab!
- Set of three
Set includes equilateral prisms of crystal clear acrylic in sizes 25 mm, 50 mm, and 100 mm.
Customers Who Bought This Also Bought