Spécifications
- Description:A. calcoaceticus
- Storage Conditions:Refrigerator
- Cat. no.:470179-054
- Packaging:Tube
- Media:Nutrient agar
- Supplier No.:WARD470179-054
- Incubation Conditions:37 °C
Spécifications
A propos de cet article
This organism is an aerobic organism that is commonly found as part of normal human intestinal flora. It is pathogenic.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Gram Stain: Negative
- Cell Morphology: Rods
- Motility: Negative
- Respiration: Aerobic
For many applications, A. calcoaceticus can be used as an alternative to Acinetobacter baumannii.
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout