Hexagel™ Electrophoresis Chamber, Edvotek
Fournisseur: Edvotek
Certificats
A propos de cet article
Run six gels simultaneously.
- Lifetime warranty
- Contoured lid for enhanced gel visualization
- Large color coded push tabs for easy lid insertion and removal
- Pour spout for buffer disposal
- User replaceable electrodes
Apparatus supports up to six groups of students. Produces excellent results in 30 to 40 minutes.
Ordering information: Includes: (6) 7×7" Gel Trays; (6) 6-tooth combs; and (12) Rubber end dams.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.