9" Giant Horseshoe Magnet
# de catalogue 470101-730
Fournisseur: H. SHAW MAGNETS LTD.
Spécifications
- Description:9" Giant Horseshoe Magnet
- Cat. no.:470101-730
- Supplier No.:50019
Spécifications
A propos de cet article
The acrylic-housed, red ceramic magnet has a built-in handle with south and north poles clearly marked.
Although powerful, this magnet is safe for little hands. Size: 23 cm.
Customers Who Bought This Also Bought