Rocks and Minerals Mounted Collection, GEOSCIENCE INDUSTRIES INC SE
# de catalogue 470112-344
Fournisseur: Geoscience Industries
Spécifications
- Description:Rocks and Minerals Collection with 50 Samples
- Cat. no.:470112-344
- Supplier No.:1050 + PKG
Spécifications
A propos de cet article
This rock and mineral collection is an engaging and economical classroom set.
- Study Guide Included
- 50 Samples Included
- Display Case: 14" x 17"
- Each Sample: 3/4"
The collection of 50 samples is contained in a 14 x 17” display box.
Each sample is approximately ¾”.
Ordering information: This collection includes a study guide and 50 samples.
Customers Who Bought This Also Bought