3B Scientific® Skin Model
# de catalogue 470004-538
Fournisseur: American 3B Scientific
Spécifications
- Description:Skin
- Height:6 cm
- No. of Keyed Items:28
- Cat. no.:470004-538
- Length:26 cm
- Width:34 cm
- Supplier No.:J10
- No. of Parts:1
Spécifications
A propos de cet article
Three Layers Of Skin From Scalp Depicted
- Shows Hairy And Hairless Skin
- 40 X Life Size
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
Delve into the inner layers of the human scalp with the colorful, raised-relief model enlarged 70X. The section through three layers reveals hair follicles with sebaceous glands, sweat glands, nerves, and vessels. The model is mounted on a base.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought