3B Scientific® Lung Model With Larynx
# de catalogue 470119-142
Fournisseur: American 3B Scientific
Spécifications
- Description:Lung With Larynx
- Height:38 cm
- No. of Keyed Items:55
- Cat. no.:470119-142
- Length:12 cm
- Width:28 cm
- Supplier No.:VC243
- No. of Parts:5
Spécifications
A propos de cet article
This detailed heart depicts the structures of the diaphragm, the lungs, heart and larynx.
- 3× life size
- Base represents the diaphragm
- 3 year manufacturers warranty
- Includes 3B smart anatomy
The following parts of the heart can be removed: Esophagus, Trachea, Superior vena cava, Aorta, Pulmonary artery stem, Both atrium walls, Both ventricle walls.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought