3B Scientific® Numbered Skulls With Open Dentition
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
This dental skull with opened mandible exposes tooth roots with vessels and nerves. The skull model also features the numbering of the cranial bones, bone components, fissures, foramina and other structures.
- Includes 3B smart anatomy
The cranial sutures are shown in color, as are the meningeal vessels and venous sinuses. Not only are important anatomical features of the skull depicted but also the open jaw gives a distinctive view of teeth roots.
The painted version includes muscle origins (red) and insertions (blue) represented on the left side of this human skull model.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought