3B Scientific® ³/₄ Size Human Anatomy Figure
# de catalogue 470029-526
Fournisseur: American 3B Scientific
Spécifications
- Description:Entire Human Anatomy Figure
- Height:55"
- No. of Keyed Items:674
- Cat. no.:470029-526
- Supplier No.:B50
- No. of Parts:45
Spécifications
A propos de cet article
Detailed Depiction Of Musculature As Well As Internal Organs
- Includes 3B Smart Anatomy
A dramatic representation of the human figure, this 3/4-size figure features 45 removable parts for in-depth study. Both arms and the left leg can be detached, as can five muscles from the left arm and eight from the leg. Removable parts also include the head, neck, calvarium showing the three-part brain, frontal plate, mammary gland, both two-part lungs, thymus, two-part heart, liver and gallbladder, duodenum and pancreas, intestines with cecal flap to show the inner structure, the front of the kidney, and four-part male and two-part female interchangeable genitals. The model is mounted on a roller stand for mobility and comes with a key identifying 674 structures.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought