3B Scientific® Classic Giant Ear
# de catalogue 470119-102
Fournisseur: American 3B Scientific
Spécifications
- Description:Classic Giant Ear
- Height:21 cm
- No. of Keyed Items:29
- Cat. no.:470119-102
- Length:36 cm
- Width:18 cm
- Supplier No.:E10
- No. of Parts:4
Spécifications
A propos de cet article
At approximately three times life-size with four removable parts, this model offers a comprehensive view of the human ear. It has representations of the outer, middle and inner ear, featuring a removable eardrum with hammer, anvil and stirrup.
- 3× life size
- Study outer, inner, or middle ear
- 3 year manufacturers warranty
- Includes 3B smart anatomy
A two-part labyrinth with cochlea and auditory/balance nerve is also shown. Model is mounted on base.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought