21st Century Balance
# de catalogue 470148-796
Fournisseur: VWR International
Spécifications
- Description:21st Century Balance
- Cat. no.:470148-796
- Supplier No.:P10854
Spécifications
A propos de cet article
This balance is the first major advancement in teaching simple machines in decades.
- Unique Features
- Internal Sliding Weight Mechanism
- Instructions Included
- Scale: 50 cm
The internal sliding weight mechanism allows users to neutralize all factors once the fulcrum and positions of force have been established.
It includes instructions and a 50 cm scale.
Customers Who Bought This Also Bought