Spécifications
- Description:Battery Holder, 2, with Plug Jacks
- Cat. no.:470148-864
- Supplier No.:S0598
Spécifications
A propos de cet article
A simple tool for communicating complex workings of electricity, this plastic battery holder is fitted with three banana plug jacks.
- Illustrate DC, series, open, and closed circuits
- Easy switch between selections of DC, 1.5 and 3 V power for your experiments
The holder fits two D-cell batteries and can be used to demonstrate circuits, Ohm's law, and the transformation of energy. Batteries not included.