Rocks, Minerals, And Gemstones of the United States, GEOSCIENCE INDUSTRIES INC SE
# de catalogue 470151-150
Fournisseur: Geoscience Industries
Spécifications
- Description:Rocks, Minerals, And Gemstones with 100 US Samples
- Cat. no.:470151-150
- Supplier No.:1100-OAK
Spécifications
A propos de cet article
Collection of rocks, minerals and gemstones typical of those found in the United States and North America.
- Includes study guide
- Big variety of specimens
- 12×20" honey oak frame
Each specimen is approximately ³/₄ to 1" and mounted in a beautiful solid honey oak frame, ready to hang on the wall.
Customers Who Bought This Also Bought