Mag Chips
# de catalogue 470110-256
Fournisseur: NADA SCIENTIFIC LTD.
Small Business Enterprise
Spécifications
- Description:“Mag Chips”
- Cat. no.:470110-256
- Supplier No.:B2637-115
Spécifications
A propos de cet article
Easier to Use and Clean Up than Iron Filings.
- They are 2mm-long pieces of zinc-plated iron wire
- When exposed to a magnetic field, they align, exhibiting the field lines clearly. 50g container
Customers Who Bought This Also Bought