Dissectible Student Leyden Jar
# de catalogue 470002-944
Fournisseur: United Scientific Supplies
Spécifications
- Description:Jar, Leyden
- Cat. no.:470002-944
- Supplier No.:LYJ001
Spécifications
A propos de cet article
Quickly demonstrate the isolation of a static charge.
- Easily assembles or disassembles
- Large electrode for attracting a charge
Learn about static electricity and the function of a dielectric with this unique device. Two aluminum jars - one with an attached aluminum electrode and one plastic jar – are used to safely collect the charge.
Customers Who Bought This Also Bought