System Labels 'Hazardous Materials', National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Place labels directly on feed hoppers, pipelines, railroad tankers, storage tanks and any other potentially hazardous equipment.
- Blank four color diamonds to identify chemicals store
Use clear labels to fill in corresponding numbers and symbols.
Certifications: Comply with NFPA 704.
Customers Who Bought This Also Bought