NMC Multiple Entry Plug Lockout
# de catalogue 94016-494
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Plug Lockout Multiple Entry
- Cat. no.:94016-494
- Supplier No.:LP550
Spécifications
A propos de cet article
Accommodates plugs up to 7.62cm (3") dia., 17.15cm (6¾") long with a maximum cord dia
of 2.54cm (1"). Accepts 220 volt and certain models of 550 volt plugs.
Customers Who Bought This Also Bought