About this item
The Barnstead MicroPure water purification systems provides up to 15L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water. All MicroPure systems include:feed water monitoring to protect your consumables, quick discconnects to make changing consumables easy, and all core consumables and tubing. MicroPure-ST systems come standard with a 6 liter integrated feed tank for labs that do not have a ready supply of pre-treated feed water.
The MicroPure systems offer highly versatile mounting options.The units can be mounted on the wall, on the bench or under the bench.
The MicroPure systems are offered in a variety of configurations to meet a wide range of applications needs.
The MicroPure systems are ideal for atomic absorption, ion chromatography or other analytical procedures.
The MicroPure UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The MicroPure UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media.
Specifications
- Catalog No:
- 89230-432
- 89230-434
- CA89230-428
- 89230-430
- W×D×H:
- 305×300×545 mm (12×11.8×21.5")
- 305×300×545 mm (12×11.8×21.5")
- 305×300×545 mm (12×11.8×21.5")
- 305×300×545 mm (12×11.8×21.5")
Specifications
Related Information
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.