Sterile Spec-Tec™ Stretch Glove Liners, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
These glove liners feature high performance fiber blended with Lycra® for stretch and reusability.
- The gloves feature good tactile sensitivity
Ambidextrous and latex free, the gloves can be washed 12 times without shrinking.
Certifications: ANSI Level 2 Cut Protection.
Specifications
- Catalog No:
- 80091-134
- 80091-132
- 80091-130
- 80091-128
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous