ContactUsLinkComponent
Labels, Blank, National Marker
Labels, Blank, National Marker
# de catalogue 94004-320
Blank Label, 2 1/2 X 2 1/8, Ps Paper, 500 Roll
Labels, Blank, National Marker
# de catalogue 94004-320
Spécifications
  • Color:
    White
  • Description:
    Blank white labels
  • Material:
    PS Paper
  • Cat. no.:
    94004-320
  • Legend:
    Blank White Labels
  • Dimensions:
    2.5x2.13"
  • Supplier No.:
    BLR01AL
Spécifications
A propos de cet article
These labels are pressure sensitive adhesive type. Easily communicate common shipping messages and information with these shipping labels.
  • Chemical resistant
  • Graffiti proof
  • Waterproof
Customers Who Bought This Also Bought