Master Lock® Universal Wall Switch Lockout, NMC
# de catalogue 94013-078
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Universal Wall Switch
- Cat. no.:94013-078
- Supplier No.:ES02
Spécifications
A propos de cet article
Designed to lockout both toggles and decora paddle wall switches
Easy to install, uses existing screws and can be used on most wall switches. Locks in either the "On" or "Off" position.
Customers Who Bought This Also Bought