Cap, Premium, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This premium cap and pour ring are designed for GL45 thread Duran bottles
Made of colorless, TPCh260 resin so that there are no leachables. Cap has high chemical resistance, and is leak-tight due to a PTFE coated silicone seal. Meets USP standards. Autoclavable.
Customers Who Bought This Also Bought