Cap, GL Thread, PTFE Membrane, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These polypropylene, blue caps have a center hole and welded-in, PTFE, inner gas permeable membrane
Ideal for media sterilization and storage. The Temperature Range (°C) for all modules is -40 to + 140°C.
Customers Who Bought This Also Bought