Adapter, Distillation, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This distillation adapter is for use with bench or pilot plant reactors.
- Center vessel collects moisture
- With bottom PTFE plug stopcock
- Fabricated from borosilicate glass
Moisture is collected in the center vessel and drained off through the bottom stopcock which is ground to accept a compression style fitting. Stopcock is a PTFE plug. Available with either one or two reinforced top standard taper outer joints, and single standard taper inner joint at bottom next to stopcock. Joint Size value represents the compression fitting joint size .
Customers Who Bought This Also Bought