Economy Wash Bottle, Ace Glass
Supplier: Ace Glass
Certificates
About this item
These wash bottles come with a top-angled spout at a money-saving price.
Bottle, tubing, and tip are made of polyethylene with a polyallomer cap and nozzle. Molded tip dispenses a uniform stream. Can be cut back to increase flow.
Customers Who Bought This Also Bought