Thermometer Adapters, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This PTFE adapter features either a Viton or Silicone O-ring for use with plain stem thermometers, gas inlet tubes, probes, etc.
- Accommodates thermometer
- With Viton or Silicone O-ring
- O-ring compression seal allows for adjustable depth positioning
- Fabricated from borosilicate glass
The 10/18 size will accommodate tubes up to 6.5 mm, and all other sizes will take tubes up to 7 mm. All adapters have an external O-ring seal (with the exception of the 10/18 size).
Ordering information: Adapter 89062-930 is designed for use with Viton O-Ring 89060-614. Adapter 89062-932 is designed for use with Viton O-Ring 89060-620. Adapter 89062-934 is designed for use with Viton O-Ring 89060-624. Adapter 89062-936 is designed for use with Viton O-Ring 89060-632. Adapter 76318-582 is designed for use with Silicone O-ring 89060-666. Adapter 76318-584 is designed for use with Silicone O-ring 89060-674.
Customers Who Bought This Also Bought