Adapter, Vacuum, Long Stem, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This apparatus is useful as a vacuum adapter or addition tube.
- Top outer [ST] joint
- Bottom inner [ST] joint
- With hose connection
- Fabricated from borosilicate glass
The stem length below the joint on this adapter is 130mm. Use with a 5/16-inch I.D. tubing, size A hose connection. Adapter has an outer standard taper joint at the top and an inner standard taper joint at the bottom.
Customers Who Bought This Also Bought