Adapter, Straight, Ace Glass Incorporated
# de catalogue 89060-454
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Straight Adapter
- Cat. no.:89060-454
- Supplier No.:7805-12
Spécifications
A propos de cet article
This adapter features two standard taper outer joints, one with a drip tube.
- Top and bottom [ST] outer joints
- Drip tube
- Fabricated from borosilicate glass
Adapter has two standard taper 14/20 outer joints, one with a 7mm drip tube.
Customers Who Bought This Also Bought