Hand Crimper, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This hand crimper is suitable for attaching aluminum seals.
This crimper features a new, ergonomic design cushioned handle that aids in reducing hand fatigue and provides a higher degree of comfort for the user. Each crimper is labeled for quick identification for seal size. Can be autoclaved.
Customers Who Bought This Also Bought