Adapter, Gas, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter features a T-Bore, 2mm stopcock at the top.
- Bottom [ST] outer joint
- Top T-Bore stopcock
- Fabricated from borosilicate glass
Adapter has a standard taper outer joint at the bottom and a T-Bore, 2mm stopcock at the top. "[ST] Joint" value represents the value of the outer [ST] joint.
Customers Who Bought This Also Bought