Adapter, Bleed Capillary, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This bleed capillary adapter is fabricated from borosilicate glass.
- Top [ST] joint
- With hose connection
- Fabricated from borosilicate glass
The length of this adapter is measured from the top of the joint to the tip. Used with a 5/16-inch I.D. tubing, size A hose connection.
Ordering information: Contact your Avantor representative for a listing of which flasks and heads fit this adapter.
Customers Who Bought This Also Bought