Adapter, Firestone, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
When inserted into a flask, this rotary evaporator “splash guard” adapter combines with flask neck to give the best protection against a splash-up.
- Top and bottom [ST] medium length joints
- Provides best splash protection
- Fabricated from borosilicate glass
The adapter's standard taper medium length joints, top and bottom, will accept full length standard taper joints. Overall height is kept to a minimum to effect best distillation.
Customers Who Bought This Also Bought