Adapter, Thermometer, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The thermometer adapter features a vacuum takeoff, or gas addition, side tube.
- Side tube accommodates thermometer
- Bottom [ST] inner joint
- Fabricated from borosilicate glass
The top center tube of this thermometer adapter is 13mm O.D., the side tube is 6mm O.D., and the inner bottom joint is an inner standard taper 24/40.
Customers Who Bought This Also Bought