Adapter, Thermocouple with 1:5 PTFE Stopcock, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Thermocouple adapter with 1:5 PTFE stopcock with hose connection.
- Stopcock with hose connection
- Fabricated from borosilicate glass
Supplied with bushing and FETFE O-ring which allows for compression seal with probes.
Customers Who Bought This Also Bought