Peligro Voltage and Electrical Signs, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Safety signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
- Ideal for indoor and outdoor labeling
- Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
- .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
- .004 PS vinyl material has pressure sensitive adhesive backing
- .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Customers Who Bought This Also Bought