Bottle, Dispensing, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Drain extension near the bottom for attaching stopcock shutoff valve. Shutoff is an 8mm bore PTFE plug stopcock with a #25 Ace-Thred at a right angle. Ace-Thred attaches to drain extension via a 89059-226 nylon bushing and a FETFE o-ring. The 9.5 and 13.5 liter sizes are conventional bottle shape; 19 liter is similar in design to a carboy. Neck is 45/50 reinforced outer standard taper joint, rather than a tooled neck for a rubber stopper. Fabricated from borosilicate glass.
This dispensing bottle has a 24 mm O.D. Necks [ST] Joints 45/50 Top
Ordering information: For full length stopper (not included), order 89062-830. For shutoff valve (not included), order 89051-642. For nylon bushing and o-ring for shutoff valve (not included), order 89059-226. All these items must be ordered separately.
Customers Who Bought This Also Bought