Short Arc Mercury Lamp Bulb, Bulbtronic©
Fournisseur: Bulbtronics
Certificats
Certificats
A propos de cet article
Microscope replacement bulb.
Customers Who Bought This Also Bought
Formaldehyde 37% in aqueous solution stabilized, VWR®, Reagent Grade
Fournisseur: VWR International
77,47 $ to 576,55 $
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
72,65 $ to 846,50 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
131,08 $ to 405,43 $