VWR® Layflat Poly Bags
Fournisseur: VWR International
Certificats
A propos de cet article
Each of VWR® imported layflat poly bags are made of 100% virgin low density polyethylene (LDPE) resin that meets FDA/USDA specifications.
- LDPE fabrication
- Extra strong bottom
- Superior clarity of siding
Manufactured with an extra strong bottom and a durable side weld for optimal strength. This premium bag features superior clarity for showing the quality of your customer's products. All listed gauges are nominal. 100 Packed. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought