NMC Single Entry Plug Lockout
# de catalogue 94016-488
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Cat. no.:94016-488
- Supplier No.:LP110
Spécifications
A propos de cet article
Accommodates plugs up to 4.45cm (1¾") dia., 8.26cm (3¼") long with a maximum cord dia
Of 1.27cm (½"). Accepts 110 volt and certain models of 220 volt plugs.
Customers Who Bought This Also Bought